MES sodium salt, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
MES-Na
0215245410
0215245480
IC15245410EA
32.29
USD
IC15245410
IC15245480
MES sodium salt, white powder
MES sodium salt
Solubility (10% aq soln): Clear Solution.
Solubility (10% aq soln): Clear Solution.
Precaución: Research Use Only (RUO).
Formula:
C₆H₁₂NNaO₄S Storage Temperature: Ambient |
MDL Number:
MFCD00065473 CAS Number: 71119-23-8 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
pH (1% aq. soln) | 8.5 - 10.4 |
Moisture Content | ≤9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...