Sodium nitroprusside dihydrate ≥99%, dark brown solid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Sodium pentacyanonitrosylferrate(III) dihydrate, Sodium nitrosopentacyanoferrate (III) dihydrate, Sodium nitroferricyanide(III) dihydrate, Disodium pentacyanonitrosylferrate (III) dihydrate, Sodium nitroferricyanide dihydrate

0215206125 0215206180 0215206190
IC15206125EA 93.13 USD
IC15206125 IC15206180 IC15206190
Sodium nitroprusside dihydrate ≥99%, dark brown solid
Sodium nitroprusside dihydrate

Sodium nitroprusside dihydrate reagent used for chromatographic detection of peptides containing cystine. Also liberates nitric oxide in biological systems.


Sodium nitroprusside dihydrate was used in the digital image analysis for the determination of products of simple colorimetric tests for amphetamine and methylamphetamine.


Sodium nitroprusside dihydrate (Disodium pentacyanonitrosylferrate(2-)dihydrate, SNP) molecule contains a ferrous ion in the center, which is surrounded by five cyanide groups and a nitrosyl group. Sodium nitroprusside has clinical applications. It acts as a rapid-acting vasodilator. It is also widely employed as nitric oxide (NO) donor in various experimental models. Two metastable states of sodium nitroprusside dihydrate, Na2[Fe(CN)5NO]·2H2O have been identified and their geometries have been studied by X-ray diffraction.

Formula: Na₂[Fe(CN)₅NO]·2H₂O
MW: 297.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149192
CAS Number: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6aa7477ccc864a4e7fd1a2ae4a29050b78d79c90&vid=6aa7477ccc864a4e7fd1a2ae4a29050b78d79c90&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR