Salinomycin Sodium Salt

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215204305 0215204325 0215204350
IC15204305EA 44.99 USD
IC15204305 IC15204325 IC15204350
Salinomycin Sodium Salt
Salinomycin sodium salt

Salinomycin sodium salt is a cationophore that promotes transport of Na+ and K+ ions through phospholipid bilayers


  • White powder


It is useful in studies concerning membrane transport of cations.


E Number E716


Soluble in ethanol

Formula: C₄₂H₆₉NaO₁₁
Melting Pt: 112.5…113.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00167109
CAS Number: 55721-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR