Safranine O, dark green powder C.I. 50240

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: CI 50240

0215204005 0215204025 0215204080
IC15204005EA 42.83 USD
IC15204005 IC15204025 IC15204080
Safranine O, dark green powder C.I. 50240
Safranine O

Safranine O is a redox and adsorption indicator.


Safranine O is a biological stain used in histology, cytology, bacteriology.


Soluble in ethanol 50% (Clear, orange-red solution with fluorescence).

Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: l max (50% ethanol) 530 ± 5 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR