D-ribulose 85, Liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215202810 0215202825 0215202880 0215202890
IC15202810EA 71.9 USD
IC15202810 IC15202825 IC15202880 IC15202890
D-ribulose 85, Liquid
D-ribulose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
CAS Number: 488-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR