Ribonucleic acid (from torula yeast)

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215202625 0215202680 0215202690
IC15202625EA 105.83 USD
IC15202625 IC15202680 IC15202690
Ribonucleic acid (from torula yeast)
Ribonucleic acid

Ribonucleic acid (RNA) from torula yeast may be used as a substrate for studying ribonuclease activities of enzymes such as ribonuclease-A, ribonuclease T1 (RNAase) and bougainvillea xbuttiana antiviral protein 1 (BBAP1).


  • Off-white powder
  • Phosphorus: <9.5%


On soluble provides colorless to light yellow to light beige solution. Contains 10 mg and 1 ml of 0.1 M acetate buffer, pH 5.0.


Precaución: Research Use Only (RUO).

Storage Temperature: Freezer
MDL Number: MFCD00132195
CAS Number: 63231-63-0

Order Now

Specification Test Results

Identity Test Passes
Source Torula Yeast
Phosphorus ≤9.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR