Pyridine, liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215198380
0215198383
IC15198380EA
74.77
USD
IC15198380
IC15198383
Pyridine, liquid
Pyridine
Soluble in water. Forms an azeotropic mixture with 3 mols water, boiling at 92-93 °C. Volatile with steam. Miscible with alcohol, ether, petroleum ether, oils and many other organic liquids. Forms salts with strong acids.
Japan Fire Code: 4.1
Formula:
C₅H₅N |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 UN: 1282 ADR: 3,II |
Specification Test Results
Synonyms | Azabenzene; Azine |
Form | Clear Liquid |
Storage | Store at Room Temperature(15-30 °C), store under nitrogen. |
pka | pKa: 5.19(Lit.) |
Refractive Index | n20/D 1.509(Lit.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...