Dipotassium hydrogen orthophosphate trihydrate ≥99%, crystals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate

02151946.1 02151946.5 0215194601 0215194610
IC151946.1EA 54.83 USD
IC151946.1 IC151946.5 IC15194601 IC15194610
Dipotassium hydrogen orthophosphate trihydrate ≥99%, crystals
Dipotassium hydrogen orthophosphate trihydrate

Soluble in water (100 mg/mL) ; Insoluble to slightly soluble in ethanol; insoluble in alcohol.


For phosphate buffers, the pH increases with decreasing temperature. Compared with a buffer at 25 °C, buffers at 4 °C will be 0.08 pH unit higher and a buffer at 37 °C will be 0.025 pH unit lower. The concentration of phosphate also influences pH. ; Typical Buffer Preparation/Formulations: 1 M Potassium Phosphate Monobasic, 1 M Potassium Phosphate Dibasic, 5 M Sodium Chloride

Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1

Order Now

Specification Test Results

Form Clear Granular Crystals
Storage Store at Room Temperature (15-30 °C).

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR