Polyvinyl alcohol, M.W. 72,000, pale yellow crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

0215194083 0215194091
IC15194083EA 78.18 USD
IC15194083 IC15194091
Polyvinyl alcohol, M.W. 72,000, pale yellow crystalline powder
Polyvinyl alcohol

Polyvinyl Alcohol(PVA) is used as a tissue stabilizer or mounting media in histochemistry. PVA has been used as a preservative. PVA forms microgels in aqueous solution.


Hydrolysis: 87.0 - 89.0%; Volatiles: ≤5.0%


Precaución: Research Use Only (RUO).

Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Identity Test Passes
Viscosity (4% aq soln) 23 - 27 cps
Hydrolysis 87.0 - 89.0%
Volatiles ≤5%
pH 4.0 - 6.0(4% aq soln)
Ash ≤0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR