Fosfomycin disodium salt

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215187605 0215187610
IC15187605EA 140.84 USD
IC15187605 IC15187610
Fosfomycin disodium salt
Fosfomycin disodium salt

Phosphomycin, also known as Fosfomycin, is an antibiotic produced by Streptomyces fradiae. Phosphomycin concentrates in kidney and bladder, reduces nephrotoxicity and ototoxicity of platinum-containing anti-tumor agents.


  • White powder


Phosphomycin disodium salt is used to treat uncomplicated urinary tract infections and to reduce nephrotoxicity and ototoxicity of platinum-containing anti-tumor agents


Phosphomycin is a phosphoenolpyruvate analog that irreversibly inhibits UDP-GlcNAc enolpyruvyl tranferase (MurA), an enzyme involved in bacterial cell wall biosynthesis. As a result, the production of N-acetylmuramic acid, an essential element of the peptidoglycan cell wall, is inhibited.


It is used for susceptibility studies of Klebsiella pneumoniae and to study in vitro susceptibility testing procedures for fosfomycin tromethamine.


Soluble in water (50 mg/mL- clear, colorless solution) and 1 M HCl (50 mg/mL)

Formula: C₃H₅Na₂O₄P
Storage Temperature: Refrigerator
MDL Number: MFCD00056853
CAS Number: 26016-99-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f046e553193f06ea963cace1d9e3fe3206c0066c&vid=f046e553193f06ea963cace1d9e3fe3206c0066c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR