Phosphoenol pyruvic acid trisodium salt hydrate

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215187225 0215187280 0215187283 0215187290 0215187291
IC15187225EA 42.01 USD
IC15187225 IC15187280 IC15187283 IC15187290 IC15187291
Phosphoenol pyruvic acid trisodium salt hydrate
Phosphoenol pyruvic acid trisodium salt hydrate

Soluble in water.

Formula: C₃H₂Na₃O₆P
MDL Number: MFCD00150737
CAS Number: 5541-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc24e394f3e8b889caa3b2da41c1226c143c9939&vid=bc24e394f3e8b889caa3b2da41c1226c143c9939&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR