p-Phenylenediamine, flakes

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 1,4-Diaminobenzene, PPD, 1,4-Phenylenediamine

0215183080 0215183083
IC15183080EA 92.97 USD
IC15183080 IC15183083
p-Phenylenediamine, flakes
p-Phenylenediamine

P-Phenylenediamine is an anti-fade reagent added to the mounting medium in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes.


Soluble in ethyl alcohol (EtOH)(50 mg/mL).


Precaución: Research Use Only (RUO). Protect from light, store under nitrogen.

Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00007901
CAS Number: 106-50-3
UN: 1673
ADR: 6.1,III

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 138 - 149 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR