Oleic acid ≥99%, clear, colorless liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (9Z)-9-Octadecenoic acid

0215178101 0215178105 0215178125
IC15178101EA 87.04 USD
IC15178101 IC15178105 IC15178125
Oleic acid ≥99%, clear, colorless liquid
Oleic acid

Oleic acid is a mono-unsaturated omega-9 fatty acid created by dehydrogenation of stearic acid. It is then further dehydrogenated and elongated to create members of the n-9 family of fatty acids.


Oleic acid is used as emollient. Small amounts of oleic acid are used as an excipient in pharmaceuticals. It is used as an emulsifying or solubilizing agent in aerosol products. Oleic acid have been shown to be important for the growth and productivity of Chinese Hamster Ovary (CHO) cells. Functions of Oleic Acid in Cell Culture Systems: Long-term energy storage: energy derived from NADPH and ATP is stored in fatty acids. Fatty acids are esterified to a glycerol backbone to form a group of compounds known as mono-, di- and tri- glycerides (neutral fats). Energy is released when fatty acids are degraded.


Soluble in methanol (40 mg/mL- clear solution), chloroform, benzene or fixed and volatile oils. Insoluble in water.


Precaución: Research Use Only (RUO).

Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Melting Pt: 13…14 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00064242
CAS Number: 112-80-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR