Nile red, dark green powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
0215174450
0215174483
0215174490
0215174410
IC15174450EA
130.46
USD
IC15174450
IC15174483
IC15174490
76102-722
Nile red, dark green powder
Nile red
Nile Red is an uncharged, hydrophobic, photostable molecule that strongly fluoresces bright red in hydrophobic environments. It is a fluorescent stain for intracellular lipid droplets.
Nile red can be used for the rapid determination of lipids by thin-layer chromatography or for staining whole tissue such as atherosclerotic blood vessels.
- Presentation: Dark Green Powder
- Soluble in ethanol (1 mg/mL) and very slightly soluble in water (< 1µg/mL).
- Storage temperature: Store at Room Temperature (15-30 °C)
Formula:
C₂₀H₁₈N₂O₂ MW: 318.38 g/mol Melting Pt: 203…205 °C (lit.) Density: 1.23 g/cm³ (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00011639 CAS Number: 7385-67-3 |
Specification Test Results
Identity Test | Passes |
UV/VIS: l Max (MeOH) | 553 ± 4, 310 ± 4 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...