ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥95%

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: NADPH tetrasodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1(4H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)oxidophosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl phosphate, Nicotinamide adenine dinucleotide phosphate tetrasodium salt, (reduced form)

0215174225 0215174250
IC15174225EA 113.73 USD
IC15174225 IC15174250
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥95%
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)

50 mg/ml, clear colorless to slightly yellow solution.


  • White Powder

Formula: C₂₁H₂₆N₇Na₄O₁₇P₃
MW: 833.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036263
CAS Number: 2646-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1c806933c09183eb0b026abc11d95aed2b16f76&vid=de428b8e8de892dc6bca9231418b66d274ba5e4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms β-NADPH; TPNH; Coenzyme II reduced form tetrasodium salt; Dihydronicotinamide-adenine dinucleotide phosphate tetrasodium salt; Triphosphopyridine nucleotide reduced tetrasodium salt
Form White Powder
Storage Store at -0°C, desiccate, protect from light, store under nitrogen.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR