Nalidixic acid sodium salt

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate

0215172401 0215172405 0215172425 0215172480
IC15172401EA 25.65 USD
IC15172401 IC15172405 IC15172425 IC15172480
Nalidixic acid sodium salt
Nalidixic acid sodium salt

Nalidixic acid sodium salt is an antibacterial agent (synthetic quinolone antibiotic), which is inhibitor of bacterial DNA polymerase (DNA gyrase) & avian myeloblastoma virus reverse transcriptase.


Nalidixic acid sodium salt is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. Inhibits nucleic acid and protein synthesis in Saccharomyces cerevisiae.


Typically used at a concentration of 15 ug/mL in molecular biology.


Soluble in water (100 mg/mL - clear, light yellow solution). Stock solutions can be aliquoted and stored at -20 °C for up to approximately 3 to 4 months.

Formula: C₁₂H₁₁N₂NaO₃
MDL Number: MFCD00064376
CAS Number: 3374-05-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db441a37fdd454b3fd02cdbfb681c2ee8d46241&vid=5db441a37fdd454b3fd02cdbfb681c2ee8d46241&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form Off-white powder
Storage Store at 0 °C,Use only in area provided with appropriate exhaust ventilation
Moisture Content ≤9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR