2-Mercaptoethanesulfonic acid sodium salt (mesna), white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
2-Mercaptoethane Sulfonic Acid Sodium Salt is a thermally stable acidic cofactor in the methyl transfer reaction of methanobacterium.
Also acts as a water-soluble disulfide reducing agent.
Soluble in water (50 mg/mL).
Precaución: Research Use Only (RUO).
Formula:
C₂H₅NaO₃S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00007535 CAS Number: 19767-45-4 |
Specification Test Results
Identity Test | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...