2,4-Dinitro-1-naphthol, dark yellow powder C.I. 10315
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
0215158805
0215158825
IC15158805EA
53.78
USD
IC15158805
IC15158825
2,4-Dinitro-1-naphthol, dark yellow powder C.I. 10315
2,4-Dinitro-1-naphthol
Soluble in methanol (0.1%-clear dark yellow solution), EGME; Less soluble in water, ethanol.
Soluble in methanol (0.1%-clear dark yellow solution), EGME; Less soluble in water, ethanol.
Formula:
C₁₀H₆N₂O₅ Melting Pt: 130…132 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00065352 CAS Number: 605-69-6 |
Specification Test Results
Identity Test | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...