Lecithin (from soy bean) ≥90%, waxy orange-yellow chunky solid, Water Dispersable Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215153925
0215153983
0215153990
IC15153925EA
228.7
USD
IC15153925
IC15153983
IC15153990
Lecithin (from soy bean) ≥90%, waxy orange-yellow chunky solid, Water Dispersable Grade
Lecithin
Lecithin is used as an emulsifier and solubilizer for drugs in an aqueous solution, also as a vehicle for liposomes.
Precaución: Research Use Only (RUO).
Storage Temperature:
Freezer |
MDL Number:
MFCD00147406 CAS Number: 8002-43-5 |
Specification Test Results
Identity Test | Passes |
Purity | ≥90% |
Source | Soybeans |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...