Heptafluorobutyric acid, anhydrous ≥97%, liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid
0215123710
0215123780
IC15123710EA
82.74
USD
IC15123710
IC15123780
Heptafluorobutyric acid, anhydrous ≥97%, liquid
Heptafluorobutyric acid
Miscible in ethanol (0.1 mL/mL, v/v - clear, colorless solution). It can be used at concentration of 0.1% in the mobile phase of an HPLC/LC-MS protocol for the detection of marine bacterioplankton siderophores.
Molarity: 7.69 M (based on pure liquid and density)(Lit.)
Formula:
C₄HF₇O₂ MW: 214.04 g/mol |
MDL Number:
MFCD00004171 CAS Number: 375-22-4 UN: 3265 ADR: 8,II |
Specification Test Results
Synonyms | Perfluorobutyric Acid; Edman Reagent No. 3; HFBA |
Form | Clear Liquid |
Storage | Store at +4 °C. |
pka | ~ 0.4 (Lit.) |
Refractive Index | n20/D 1.3(lit.) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...