HEPPSO(β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulfonic acid) ≥99%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-hydroxypropanesulphonic acid

0215123610 0215123650 0215123680
IC15123610EA 79.61 USD
IC15123610 IC15123650 IC15123680
HEPPSO(β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulfonic acid) ≥99%, white powder
HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid)

Used as a buffer in biological research. HEPPSO is an efficient separator in protein resolution by IEF.


Soluble in water (100 mg/mL - clear, colorless solution).


Precaución: Research Use Only (RUO).

Formula: C₉H₂₀N₂O₅S
MW: 268.3305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038351
CAS Number: 68399-78-0

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
pH (1% aq soln) Record Result

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR