Guanidinium hydrochloride 99, powder_crystalline_white Optical Density Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
Guanidine Hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of proteins.
This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies and be used for the recovery of periplasmic proteins. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. It is also used in the isolation of RNA to dissociate the nucleoprotein into its nucleic acid and protein moieties. It is an inhibitor of RNase.
Formula:
NH₂C(=NH)NH₂·HCl MW: 95.53 g/mol Melting Pt: 185 |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 Merck Index: 13,04578 |
|
Specification Test Results
Identity Test | Passes |
Purity | ~99% |
FTIR | Conforms to Standard |
Melting Point | 180 - 190 °C |
OD260 (6.0 M aq soln) | ≤0.03 |
Solubility (6.0 M aq soln) C | Clear, Colorless Solution |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...