L-Glutamic Dehydrogenase, MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215118950 0215118980 0215118990
IC15118950EA 106.65 USD
IC15118950 IC15118980 IC15118990
L-Glutamic Dehydrogenase, MP Biomedicals
Enzymes

For Continuous Spectrophotometric Rate Determination method: 0.3 to 0.6 unit/ml of L-Glutamic dehydrogenase in cold Triethanolamine buffer.


One unit will cause the transformation of 1.0 µmole of alpha-ketoglutarate per minute at pH 7.3 and 25 °C.


LDH ≤0.01% ; MDH ≤0.01% ; Inhibitor: Dimethyl dicarbonate.

Order Now

SPECIFICATIONS

Enzyme Name L-Glutamic Dehydrogenase
Environmentally Preferable
Source
Species
CAS No. 9029-12-3
Form Suspension
Grade USP

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR