Evans blue ~85%, black powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
0215110805
0215110850
0215110810
0215110880
IC15110805EA
55.12
USD
IC15110805
IC15110850
76102-702
76102-704
Evans blue ~85%, black powder
Evans blue
Evans Blue used in double-labeling procedure for studying axonal branching.
Dye content: ~85%
Precaución: Research Use Only (RUO).
Formula:
C₃₄H₂₈N₆O₁₄S₄ Storage Temperature: Ambient |
MDL Number:
MFCD00004021 CAS Number: 314-13-6 |
|
Specification Test Results
Identity Test | Passes |
UV/VIS: λmax (H₂O) | 601 - 611nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...