n-Dodecyl-β-D-maltoside ~97%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research.
It has also been used for the isolation of the mitochondrial cytochrome c oxidase.
Soluble in water (100 mg/mL) - clear to very slightly hazy, colorless solution.
Precaución: Research Use Only (RUO).
Formula:
C₂₄H₄₆O₁₁ MW: 510.6 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00043012 CAS Number: 69227-93-6 |
Specification Test Results
Identity Test | Passes |
Purity | ~97% |
FTIR | Conforms to Reference |
Optical Rotation | (c=1, methanol) +47 ° ± 5 ° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...