Dimidium bromide, dark reddish purple powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide

0215095880 0215095890
IC15095880EA 72.01 USD
IC15095880 IC15095890
Dimidium bromide, dark reddish purple powder
Dimidium bromide

Dimidium Bromide is used as fluorescent probe for nucleic acids and as interchelating agent.


  • Presentation: Dark Reddish Purple Powder
  • Soluble in water (4 mg/mL - clear to hazy orangish-red solution).
  • Storage temperature: Store at Room temperature (15 - 30 °C).

Formula: C₂₀H₁₈BrN₃
MW: 380.29 g/mol
Melting Pt: 243…248 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011757
CAS Number: 518-67-2
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e669239c79726ce91f44ef372cbfed426e90f93b&vid=74b03720866852364cb9050de861a4f7c0e5e57b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR