3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) (synthetic), powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 3,3',4,4'-Biphenyltetramine tetrahydrochloride, DAB HCl

0215082501 0215082505 0215082510
IC15082501EA 88.3 USD
IC15082501 IC15082505 IC15082510
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) (synthetic), powder
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl)

Easily soluble in water. The aqueous solution is unstable and can be used only for a few days even if it is kept in a refrigerator. 1% aqueous solution is dark and turns reddish when exposed to light.


Color changes to gray or gray brown when exposed to the direct sun light.

Formula: C₁₂H₁₈Cl₄N₄
MW: 360.11 g/mol
MDL Number: MFCD00012969
CAS Number: 7411-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR