Crystal ponceau 6R, red powder C.I. 16250
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215073201
0215073205
IC15073201EA
52.07
USD
IC15073201
IC15073205
Crystal ponceau 6R, red powder C.I. 16250
Crystal ponceau 6R
Crystal Ponceau 6R is a biological protein stain, only slightly staining cellulose used in microscopy.
Crystal Ponceau 6R is useful in histology for staining fibrin in conjunction with MSB (Martius, Scarlet and Blue).
Soluble in water (10 mg/ml), alcohol, xylene, cellosolve ethylene, acetone, glycerol, dioxane, etc.
Formula:
C₂₀H₁₂N₂Na₂O₇S₂ Melting Pt: >300 °C (lit.) |
CAS Number:
2766-77-0 |
Specification Test Results
Identity Test | Passes |
UV/Vis: l max (water) | 511 ± 6 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...