Collagenase, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215070501
0215070505
IC15070501EA
796.94
USD
IC15070501
IC15070505
Collagenase, MP Biomedicals
Enzymes
Collagenase can be solubilized in calcium-free solutions such as Hank's Balanced Salts or Earles Balanced Salt Solution.
Unit definition: One unit liberates one µmole of L-leucine equivalents from collagen in 5 hours at 37 °C, pH 7.5.