Cinnamaldehyde ≥99%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215069080 0215069090 0215069091
IC15069080EA 117.92 USD
IC15069080 IC15069090 76102-690
Cinnamaldehyde ≥99%, liquid
Cinnamaldehyde

FEMA Number 2286, Flavis number 5.014 , Council of Europe no. 102c, Organoleptic: cinnamon; clove; spicy, allergen: no known allergens.


  • Presentation: Liquid
  • Vapor Pressure: 2.25 hPa at 20 °C (Lit.)
  • Refractive Index: n20/D 1.621(Lit.)
  • Solubility: Misc with alcohol, ether, chloroform, oils.
  • Storage temperature: +4°C, store under nitrogen


FEMA Number 2286, Flavis number 5.014 , Council of Europe no. 102c, Organoleptic: cinnamon; clove; spicy, allergen: no known allergens

Formula: C₉H₈O
Boiling Pt: 246 °C (1014 hPa) (lit.)
Melting Pt: –7.5 °C (lit.)
Density: 1.0497 g/cm³ (20 °C) (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00007000
CAS Number: 104-55-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c5eda5095a38fc24e404bd71266daaac8b6ac13&vid=5c5eda5095a38fc24e404bd71266daaac8b6ac13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR