Chlorogenic acid, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-O-Caffeoylquinic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid

0215061801 0215061805
IC15061801EA 161.96 USD
IC15061801 IC15061805
Chlorogenic acid, powder
Chlorogenic acid

Soluble in water or ethanol (25 mg/mL - clear to hazy, yellow solution).

Formula: C₁₆H₁₈O₉
MW: 354.31 g/mol
MDL Number: MFCD00003862
CAS Number: 327-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c68d7c76f08a0313248483063ae18ee72e6cc9a1&vid=bfa3a4c2cb986d55289b6ae698e6b65be9c3ddf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form Off-white Powder
Storage Store at Room Temperature (15-30 °C).
Source Green coffee bean or coffee seeds

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR