Cefadroxil, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Cefadroxil monohydrate, (6R,7R)-7-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

0215058101 0215058105
IC15058101EA 58.09 USD
IC15058101 IC15058105
Cefadroxil, white powder
Cefadroxil

Antibiotic. Cefadroxil was used to investigate its interaction with hPEPT1 and dipeptide Gly-Sar on basis of steady-state and presteady-state analysis thus testing the hypothesis that variations in drug response among individuals are caused by alterations in the genes involved in drug absorption.


Slightly soluble in water; practically insoluble in alcohol, chloroform, ether.


Precaución: Research Use Only (RUO). Protect from light.

Formula: C₁₆H₁₇N₃O₅S
MW: 363.388 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00865091
CAS Number: 50370-12-2

Order Now

Specification Test Results

Identity Test Passes
Potency Report Result (µg/mg)
Melting Point 190 - 205 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR