Bromothymol blue sodium salt, powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate
0215052405
0215052425
IC15052405EA
56.93
USD
IC15052405
IC15052425
Bromothymol blue sodium salt, powder
Bromothymol blue sodium salt
Formula:
C₂₇H₂₇Br₂NaO₅S MW: 646.37 g/mol |
MDL Number:
MFCD00077263 CAS Number: 34722-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...