4-Aminopyridine ∼98%, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Pyridin-4-amine, 4-Pyridylamine

0215036605 0215036680
IC15036605EA 53.52 USD
IC15036605 IC15036680
4-Aminopyridine ∼98%, crystalline powder
4-Aminopyridine

Soluble in water (50 mg/mL - clear and colorless), 100% ethanol, methanol (slightly), alcohol, and DMSO (100 mM), slightly soluble in benzene, ether; very slighly soluble in ligroin.

Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 273
Flash Pt: 156
MDL Number: MFCD00006439
CAS Number: 504-24-5
UN: 2671
ADR: 6.1 II
Merck Index: 13,03964

Order Now

Specification Test Results

Synonyms Amino-4-Pyridine; γ-Aminopyridine; 4-Pyridinamine, 4-Pyridylamine; Fampridine; 4AP
Form White Crystalline Powder
Storage Store at Room Temperature (15-30 °C).
Extinction Coefficient EmM = 16.6 (0.1 M HCl), 17.4 (0.1 M HCl)(Lit.)
pH 10.5 - 11.5 (1% aq. soln.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR