Amikacin

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02150342.2 0215034201 0215034205
IC150342.2EA 75.72 USD
IC150342.2 IC15034201 IC15034205
Amikacin
Amikacin

Amikacin is an aminoglycoside antibiotic used to treat different types of bacterial infections


  • Presentation: White Powder
  • Optical Rotation: +100.5°
  • Sparingly soluble in water
  • Loss on drying: <9%
  • pH: 9.5 - 11.5 (1% aqueous solution)


Amikacin is most often used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Enterobacter.


Amikacin prevents bacterial protein synthesis by binding to the 30S ribosome subunit and inducing mRNA misreading. It interferes with the translocation of tRNA from the A-site to the P-site.


Amikacin works by binding to the bacterial 30S ribosomal subunit, causing misreading of mRNA and leaving the bacterium unable to synthesize proteins vital to its growth. Amikacin must be given by the intravenous or intramuscular route. There is no oral form available as amikacin is not absorbed orally.


Sparingly soluble in water

Formula: C₂₂H₄₃N₅O₁₃
MW: 585.61 g/mol
Boiling Pt: 981.8 °C (760 mmHg)
Density: 1.6 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00883675
CAS Number: 37517-28-5

Order Now

Specification Test Results

pH (1% aq soln) 9.5-11.5
Loss on Drying ≤9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR