Acetoacetyl coenzyme a trisodium salt tetrahydrate ≥90%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215022401 0215022425
IC15022401EA 64.82 USD
IC15022401 IC15022425
Acetoacetyl coenzyme a trisodium salt tetrahydrate ≥90%, Powder
Acetoacetyl coenzyme a trisodium salt tetrahydrate

Soluble in water (50 mg/ mL-clear colorless to faint yellow solution).

Formula: C₂₅H₃₇N₇O₁₈P₃SNa₃·4H₂O
CAS Number: 102029-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR