2,6-Dichlorophenolindophenol sodium salt, dark green powder indicator
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide, Tillmans' reagent, 2,6-Dichloroindophenol sodium, 2,6-Dichloroindophenol sodium salt, 2,6-Dichloroindophenol sodium derivative
Indicator for vitamin C.
2,6-dichlorophenolindophenol sodium salt is a redox dye commonly used as a monitor of the light reactions in photosynthesis because it is an electron acceptor that is blue when oxidized and colorless when reduced. It is part of the Hill reagents family. DCPIP can also be used as an indicator for Vitamin C. If vitamin C, which is a good reducing agent, is present, the blue dye, which turns pink in acid conditions, is reduced to a colorless compound by ascorbic acid.
Freely soluble in water, alcohol; soluble in 0.1%(50% ethanol-clear blue solution), (ethanol + 1 N 1mL Na₂CO₃).
Precaución: Research Use Only (RUO).
Formula:
C₁₂H₆Cl₂NNaO₂ MW: 290.08 g/mol Storage Temperature: Ambient |
CAS Number:
620-45-1 |
Specification Test Results
Identity Test | Passes |
UV/VIS: λ max (water) | 604 ± 5 nm |
Moisture | ≤12% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...