Ammonium hydrogen carbonate, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Ammonium bicarbonate
Ammonium bicarbonate is used in the manufacture of porous plastics, ceramics, dyes, and pigments, fire extinguishers. It has been used in the preparation and isolation of neoglycoprotein conjugates.
It is a good buffer for applications such as lyophilization and matrix assisted laser desorption and ionization (MALDI) mass spectrometry. It is utilized for the in-gel digestion of proteins by trypsin and in the MALDI mass spectrometric analysis of proteins.
Soluble in water, decomposed by hot water 1 g dissolves in 10 mL glycerol; insoluble in alcohol and acetone.
Precaución: Research Use Only (RUO).
Formula:
NH₄HCO₃ MW: 79.06 g/mol Melting Pt: 35 Storage Temperature: Ambient |
MDL Number:
MFCD00012138 CAS Number: 1066-33-7 Merck Index: 13,00497 |
Specification Test Results
Identity Test | Passes |
pH (1% aq soln) | 7.6 - 8.1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...