5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥99%, off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate

0215004280 0215004283
IC15004280EA 102.54 USD
IC15004280 IC15004283
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥99%, off-white powder
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)

A chromogenic substrate for alkaline phosphatase.


For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry. BCIP disodium salt is soluble in water. It may be used to prepare a stock solution, which in combination with NBT and a reaction buffer, form a substrate solution for alkaline phosphatase. This substrate system, when incubated with alkaline phosphatase, produces an insoluble NBT diformazan product that is easily observable with its purple color.


Disodium salt is soluble in water (50 mg/ml); insoluble in dimethylformamide.

Formula: C₈H₄BrClNNa₂O₄P
MW: 370.43 g/mol
Melting Pt: >300 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00036757
CAS Number: 102185-33-1

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference
Loss on Drying ≤10%
UV/VIS: I max (water) 291 ± 3nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR