Kanamycin sulphate ∼735 μg/mg (from Streptomyces kanamyceticus), white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
Kanamycin Monosulfate is an aminoglycoside bacteriocidal antibiotic which acts as an inhibitor of protein biosynthesis by producing a misreading of the 70s-ribosome.
Kanamycin Monosulfate used in biotechnology applications to inhibit protein synthesis. It is also used as a selection agent for cells transformed with kanamycin B (neoR, kanR) resistance gene.
Mode of Action: Binds to 70S ribosomal subunit; inhibits translocation; elicits miscoding. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria, and mycoplasma.
- Presentation: White Powder
- Soluble in water(10-50 mg/mL).
- Solutions should be filter sterilized. Store in small aliquots at appropriate temperature until use.
- Storage temperature: Store at Room Temperature (15-30 °C)
- Appearance: White
Mode of Action: Binds to 70S ribosomal subunit; inhibits translocation; elicits miscoding. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria, and mycoplasma.
Formula:
C₁₈H₃₈N₄O₁₅S MW: 582.6 g/mol Melting Pt: >250 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 |
Specification Test Results
Identity Test | Passes |
Source | Streptomyces kanamycetikus |
Potency | ~735 μg/mg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...