L-Tyrosine disodium salt hydrate, cream colored powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
(S-3-(4-Hydroxyphenyl)alanine disodium salt hydrate, Disodium 2-Amino-3-(4-oxidophenyl) propanoate hydrate, L-3-(4-Hydroxyphenyl)alanine disodium salt hydrate, H-Tyr-OH.2Na hydrate
0210557325
0210557390
IC10557325EA
67.17
USD
IC10557325
IC10557390
L-Tyrosine disodium salt hydrate, cream colored powder
L(-)-Tyrosine disodium salt
Soluble in water (100 mg/mL - clear, colorless to slightly yellow solution).
Soluble in water (100 mg/mL - clear, colorless to slightly yellow solution).
Formula:
C₉H₉NO₃Na₂·xH₂O MW: 225.2 g/mol (anhy) Storage Temperature: Refrigerator |
MDL Number:
MFCD08276422 CAS Number: 122666-87-9 |
Specification Test Results
Synonyms | L-3-(4-Hydroxyphenyl)alanine disodium salt; disodium 2-Amino-3-(4-oxidophenyl) propanoate |
Form | Cream-colored Powder |
Storage | Store at +4 °C. |
Optical Rotation | -5° to -15°(c = 2, water) |
pH | 10.65 - 12.65(2% aq soln) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...