Hypoxanthine disodium salt ≥98.0% (dried basis), off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 1,7-Dihydro-6H-purin-6-one disodium salt, 6-Hydroxypurine disodium salt

0210545105 0210545125
IC10545105EA 86.42 USD
IC10545105 IC10545125
Hypoxanthine disodium salt ≥98.0% (dried basis), off-white powder
Hypoxanthine disodium salt

Hypoxanthine is a nutrient additive for a variety of cell culture applications involving bacterial, parasite (Plasmodium falciparum) and animal cells. Hypoxanthine is a component of selection media used in hybridoma technologies.


  • Nutrient additive for cell culture


Soluble in water (30 mg/mL)clear, colorless solution.


Precaución: Research Use Only (RUO).

Formula: C₅H₂N₄Na₂O
MW: 180.08 g/mol
Melting Pt: >300 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00005725
CAS Number: 68-94-0

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Reference
UV/Vis: l max (0.1 N HCl ) 246 - 249 nm
OD250/OD260 1.40 - 1.55
OD280/OD260 0.01 - 0.15
pH (1% aq soln) 11.5 - 12.5
Assay (on dried basis) ≥98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR