Coenzyme A trihydrate

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: CoA trihydrate

0210480910 0210480950 0210480980
IC10480910EA 77.66 USD
IC10480910 IC10480950 IC10480980
Coenzyme A trihydrate
Coenzyme A trihydrate

Coenzyme A (CoA) is an essential metabolic cofactor synthesized from cysteine, pantothenate, and ATP. CoA plays important roles in many metabolic pathways, including the tricarboxylic acid cycle, and the synthesis and oxidation of fatty acids. One of the main functions of CoA is the carrying and transfer of acyl groups. Acylated deriviates, for example acetyl-CoA, are critical intermediates in many metabolic reactions. CoA levels can be altered during starvation, and in conditions such as cancer, diabetes, and alcoholism.


Coenzyme A facilitates removal of lipid peroxides by increasing mobilization of fatty acids, and promote repair of plasma membranes by activating phospholipid synthesis.


Soluble in water (50 mg/mL - clear, colorless to faint yellow solution).


Coenzyme A facilitates removal of lipid peroxides by increasing mobilization of fatty acids, and promote repair of plasma membranes by activating phospholipid synthesis.

Formula: C₂₁H₃₆N₇O₁₆P₃S·3H₂O
MW: 821.58 g/mol
MDL Number: MFCD00065465
CAS Number: 85-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbeaf4ec4da511023dc41cebc14f15a91e8aca32&vid=73d84bb20ab28926f6495d4b45d65ebe56d3bb9f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR