Peptone, Powder, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0210480801 0210480805 0210480810
IC10480801EA 434.35 USD
IC10480801 IC10480805 IC10480810
Peptone, Powder, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Peptone is used in culture media in bacteriology, the growth requirements of most non fastidious organisms will be fulfilled by the range of amino acids, peptides and proteoses in this mixture.