Caffeic acid ≥98%, yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid

0210479701 0210479705 0210479725
IC10479701EA 39.08 USD
IC10479701 IC10479705 IC10479725
Caffeic acid ≥98%, yellow powder
Caffeic acid

Caffeic acid is used as an anti-oxidant.


Inhibits the synthesis of leukotrienes that are involved in immunoregulation, inflammation and allergy. Inhibits Cu2+-induced LDL oxidation.


Soluble in ethanol (50 mg/mL)(Lit.).


Caffeic acid is used as an anti-oxidant.

Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00004392
CAS Number: 331-39-5

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR