Yeast Extract, Yellow-Tan Powder, MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210330380 0210330390 0210330391
IC10330380EA 55.88 USD
IC10330380 IC10330390 IC10330391
Yeast Extract, Yellow-Tan Powder, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology

Yeast Extract Powder is a vacuum-dried extract concentrate of Baker's yeast containing the B-complex factors of approximately three times the weight of ordinary dry yeast. This product is a vacuum dried autolyzed yeast extract produced from 100% Baker's yeast. It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast. Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.
Yeast Extract Powder is a highly nutritive medium recommended for plate count of microorganisms in water. It is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications. Also suitable for alternative to beef extract.


  • Suitable for alternative to beef extract
  • Presentation: Yellow-Tan Powder
  • pH: 5-5.5 (aq.soln) (Lit.)
  • Storage temperature: Room Temperature, desiccate
  • Solubility (5g/100ml): Clear
  • Moisture: <5%
  • Mold Count: < 50/gram
  • Standard Plate Count: < 50,000/gram
  • Salmonella: Negative


Yeast Extract Powder is a highly nutritive medium recommended for plate count of microorganisms in water. It is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications. Also suitable for alternative to beef extract,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR