Vitamin B12 (cyanocobalamin) 0.1% in mannitol , 96%, off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210327810 0210327825 0210327880
IC10327810EA 82.01 USD
IC10327810 IC10327825 IC10327880
Vitamin B12 (cyanocobalamin) 0.1% in mannitol , 96%, off-white powder
Vitamin B12 (cyanocobalamin)

Vitamin B12, 0.1% in Mannitol is 96% pure vitamin B12 tritrated onto mannitol at 0.1% concentration.

Formula: C₆₃H₈₈CoN₁₄O₁₄P
MDL Number: MFCD00151092
CAS Number: 68-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR