Vitamin A palmitate in corn oil, >1,000,000 IU/g, clear yellow liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Retinyl palmitate, Retinol palmitate, all-trans-Retinyl palmitate

0210326325 0210326380 0210326305
IC10326325EA 220.42 USD
IC10326325 IC10326380 76204-012
Vitamin A palmitate in corn oil, >1,000,000 IU/g, clear yellow liquid
Vitamin A palmitate

  • Presentation : Clear Yellow Liquid


Storage: under nitrogen.

Formula: C₃₆H₆₀O₂
MW: 524.87 g/mol
Melting Pt: 28...29 °C (lit.)
Flash Pt: 194 °C (381.2 °F) (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00019414
CAS Number: 79-81-2

Order Now

Specification Test Results

Identity Test Passes
Activity >1,000,000 IU/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR