Uridine ≥99%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Uridine is one of the four basic components of ribonucleic acid (RNA); the other three are adenosine, guanosine, and cytidine. Upon digestion of foods containing RNA, uridine is released from RNA and is absorbed intact in the gut. Uridine plays a role in the glycolysis pathway of galactose.
Uridine is used in treatment of bipolar disorder associated with mood swings.Uridine is used in metabolic labeling and analysis of rRNA processing.
- Presentation: White Powder
- Soluble in 0.1 N HCl (50 mg/mL - clear, colorless solution).
- Storage temperature: Room Temperature
Uridine is one of the four basic components of ribonucleic acid (RNA); the other three are adenosine, guanosine, and cytidine. Upon digestion of foods containing RNA, uridine is released from RNA and is absorbed intact in the gut. Uridine plays a role in the glycolysis pathway of galactose.
Formula:
C₉H₁₂N₂O₆ Melting Pt: 160…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006526 CAS Number: 58-96-8 |
Specification Test Results
Identity Test | Passes |
Purity | ~99% |
FTIR | Conforms to Standard |
Melting Point | 160-171°C |
UV/Vis: l max (water) | 260-263nm |
OD250/OD260 | 0.72-0.77 |
OD280/OD260 | 0.32-0.38 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...