Uracil, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Pyrimidine-2,4(1H,3H)-dione
0210320425
0210320480
0210320491
IC10320425EA
69.9
USD
IC10320425
IC10320480
IC10320491
Uracil, white powder
Uracil
Uracil is a pyrimidine derivative.
Used in drug delivery and in pharmaceuticals. In addition to this, it is used in the synthesis of caffeine and in the determination of microbial contamination of tomatoes.
Soluble in NaOH (40 mg/mL - clear, colorless solution, in hot water, ammonia water and other alkalies; slightly soluble in cold water; almost insoluble in alcohol and ether.
Formula:
C₄H₄N₂O₂ MW: 112.09 g/mol Boiling Pt: 379 Melting Pt: 194 Storage Temperature: Ambient |
MDL Number:
MFCD00006016 CAS Number: 66-22-8 Merck Index: 12,09985 |
Specification Test Results
Synonyms | 2,4-Dihydroxypyrimidine; 2,4-Pyrimidinediol; 2,4(1H,3H)-Pyrimidinedione |
Form | White Powder |
Storage | Store at Room Temperature (15-30 °C). |
Extinction Coefficient | EmM 7.9 - 8.2 (in pH 7.0)(Lit.) |
pH | Neutral to Litmus Paper (4% aq susp) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...